VWR® Primaide™ System Manager
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Primaide™
903-0743EA 0 DKK
903-0743 903-0708
VWR® Primaide™ System Manager
Software Laboratoriesoftware
Chromatography Data System (CDS) for control of the VWR Hitachi Primaide HPLC Systems.
Functions for: Method creation, data acquisition, chromatograph data processing, DAD data processing and report generation.
PC requirements:
OS: Windows 7 32/64 bit or Windows 10 64 bit
CPU: Intel® Core2 Duo 2 GHz or higher
RAM: 2 GB or more required
Hard disk: At least 2 GB of free space on a partition containing the Windows® 7 system files
Display: Capable of supporting 1280×1024 pixels (256 colors or higher)
Advarsel: Not 21 CFR Part 11 Compliant, therefore not suitable for GMP/GLP environments. If 21 CFR Part 11 compliant software is required please check DataApex ClarityVA or Agilent Openlab in our webshop.