Stand magnifier
Leverandør: Eschenbach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ESCH2626EA 638 DKK
ESCH2626
Stand magnifier
Aspheric magnifier with a modern white plastic frame and an easy access to view material.
  • PXM® lightweight lens
  • Ceratec® scratch resistant coating
  • Open transparent plastic base for writing