Freezers, NoFrost, forced air cooling
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NoFrost
471-0861EA 0 DKK
471-0861 471-0862 LIEBGGV5060X LIEBGGV5010X 471-0863 471-0864 LIEBGGV5860X
Freezers, NoFrost, forced air cooling
Frysere Laboratorie-/frysere til almindelige formål
NoFrost freestanding freezers constructed of stainless steel. It features forced air cooling that maintains an even refrigeration temperature throughout the interior. The integrated temperature alarm indicates if the lower or upper alarm thresholds have been breached. It uses both acoustic and visual signals to provide maximum security for goods that have been stored.
  • Height-adjustable feet ensures easy and convenient cleaning under the appliance
  • Stylish swingline design
  • Equipped with plastic coated wire shelves and ergonomic slimline stainless steel door handle
Temperature range: −14 to −28 °C