6-Hydroxynicotinsyre
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC15521080EA
1020
DKK
EHERC15521080
6-Hydroxynicotinsyre
6-Hydroxynicotinsyre
Formel:
C₆H₅NO₃ Molekylvægt: 139,11 g/mol Smeltepunkt: 304…306 °C |
MDL Number:
MFCD00006277 CAS-nummer: 5006-66-6 EINECS: 225-682-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...