VWR® Safety Containers, Leakproof
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
720-1991EA 2270 DKK
720-1991
VWR® Safety Containers, Leakproof
Bærere Flaskebærere
Transparent container for the transport of biological samples (up to 180 pieces).
  • Shock resistant and hermetic, leakproof
  • PC hooks for a quick and safe closure
  • Emergency procedures and safety measures are indelibly printed
  • Transparent to ensure continuous monitoring of the transported samples
  • Autoclavable and chemically sanitisable to avoid cross-contamination
Bestillingsoplysninger: These containers are supplied without handle which can be ordered separately.