4-Chlorophenyl acetate

Leverandør: EHRENSTORFER GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

EHERC11473200EA 1040 DKK
EHERC11473200
4-Chlorophenyl acetate
4-Chlorophenyl acetate
Formel: C₈H₇ClO₂
CAS-nummer: 876-27-7
EINECS: 000-000-0
UN nummer: 0000

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR