Phenicarbazid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAP0227-25GEA 435 DKK
TCIAP0227-25G
Phenicarbazid ≥98.0% (by HPLC, titration analysis)
Phenicarbazid
Formel: C₇H₉N₃O
Molekylvægt: 151,17 g/mol
Smeltepunkt: 172 °C
MDL Number: MFCD00007942
CAS-nummer: 103-03-7
EINECS: 203-072-3
Merck Index: 12,07378
https://dk.vwr.com/stibo/web/std.lang.all/09/59/28990959.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR