Illuminated portable microscopes
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
630-1622EA 0 DKK
VWRI630-1622 VWRI630-1621
Illuminated portable microscopes
These handheld units are suitable for use in the field.
  • Adjustable light source onto the examination area
  • Focus control knob for an accurate view
  • Zoom microscope with a snap-on microscope slide holder
Leveringsinformation: Supplied with AA batteries and a vinyl carrying case.