Temporary horizontal lifeline, Miller®
Leverandør: Honeywell Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Miller®
118-0439EA 4200 DKK
118-0439
Temporary horizontal lifeline, Miller®
Faldbeskyttelse Faldværn
Durable braided rope with two galvanised connectors and length adjuster. Ideal for building and construction as well as for industrial applications.
  • Fast and easy to install
  • Fixing to either permanent anchorage points or temporary webbing anchorage slings
  • For use by one person only
  • Supplied with two 1,5 m webbing anchorage slings
EN 362 (04), EN795(b); Cat. III
Certificeringer: EN 362 (04), EN 795(b); Cat. III