Hotplate, SLH
Leverandør: SI Analytics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SCOI285416420EA 4060 DKK
SCOI285416420
Hotplate, SLH
Varmeapparater Varmeplader
The compact design and small footprint of this hotplate makes it ideal for use on crowded laboratory tables or under fume hoods. Compatible with most standard size borosilicate bottles, beakers and flasks.
  • Ceramic coated stainless steel plate highly resistant against strong acids and bases
  • Speed control knob for a range of 60 to 1500 rpm
The powder coated aluminium cast housing is heat resitant and can withstand heavy laboratory use. The drive motor is equipped with ball bearings for smooth running and a long service life. Two control LEDs on the front plate light up when heating and stirring function are on.