Dihydroergotamine mesylate

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

EDQMD1950000EA 1770 DKK
EDQMD1950000
Dihydroergotamine mesylate
Dihydroergotamine mesylate
Formel: C₃₄H₄₁N₅O₈S
Molekylvægt: 679,78 g/mol
MDL Number: MFCD00058615
CAS-nummer: 6190-39-2
EINECS: 228-235-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR