Staining trough
Leverandør: Hecht
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HECH41202014EA 939 DKK
HECH41202014 631-0342 HECH41201042 HECH41202042
Staining trough
Dåser Farvekrukker
Staining system contains staining jar with ground-in cover with knob made of glass, rack and handle made from stainless steel.
Suitable for 26 mm wide microscopic slides. Ideal for serial stainings, especially for the method according to Papanicolaou.