Automatic and manual rope grab, MF51 / MF52
Leverandør: Miller by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DALL1002876EA 1130 DKK
DALL1002876 DALL1002875
Automatic and manual rope grab, MF51 / MF52
These light, industrial proven, stainless steel automatic and manual rope grabs are suitable for vertical, horizontal and sloping applications.
  • Automatic locking for vertical use
  • Rope glides freely over the line without manual intervention
  • Manual position for horizontal work