Adapter-S, KF, for reaction vessels with cooling jacket
Leverandør: Witeg
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WITG0.998.129EA 690 DKK
WITG0.998.129
Adapter-S, KF, for reaction vessels with cooling jacket
Adapter Adaptere med kulslipning
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
S-adapters for thermostatic reaction vessels with cooling jacket.