Puromycin dihydrochloride
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
HYCLSV30075.01EA 0 DKK
HYCLSV30075.01
Puromycin dihydrochloride
Puromycin dihydrochloride
Formel: C₂₂H₂₉N₇O₅·2HCl
Molekylvægt: 544,44 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00012691
CAS-nummer: 58-58-2
EINECS: 200-387-8