VWR®, Human ribonuclease inhibitor (from placenta)
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
E633-2KUEA 2200 DKK
E633-2KU
VWR®, Human ribonuclease inhibitor (from placenta)
Proteiner
RNase inhibitor is used to inhibit the activity of RNases in reaction mixtures for cDNA synthesis and in vitro transcription, as well as for long-term storage of valuable RNA samples. A unit of this protein will inhibit 50% of the activity of 5 ng of RNase A.
Human placental source, 40 KU/ml
Activity @ 25 °C: 10 U/ul
DNase (endonuclease): None
DNase (exonuclease): None