m-Cresol purple, Supelco®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M-Clarity™ quality level = MQ200
1.05228.0005EA
1090
DKK
1.05228.0005
m-Cresol purple, Supelco®
m-Cresol purple
Formel:
C₂₁H₁₈O₅S Molekylvægt: 382,43 g/mol |
MDL Number:
MFCD00005871 CAS-nummer: 2303-01-7 EINECS: 218-960-6 |
Specifikationer Testresultater
Solubility | (20 °C) slightly soluble |
Molar Mass | 382.43 g/mol |
Bulk Density | 480 kg/m3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...