Temperature indicators, 3M™ Freeze Watch™
Leverandør: 3M Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Freeze Watch™
MMME9805FWEA 186 DKK
MMME9805FW MMME9800FW
Temperature indicators, 3M™ Freeze Watch™
Temperaturindikatorstrips
Freeze watch indicators consist of a highly sensitive indicating liquid inside a specially designed ampoule to monitor exposure of temperature-sensitive products to freezing temperatures.
  • Easily-interpreted visual results
  • Self-adhesive backing sticks securely to most exterior packaging materials