Agarose 3:1 HRB™ for biotechnology
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E776-100GEA 2690 DKK
VWRCE776-100G
Agarose 3:1 HRB™ for biotechnology
Agarose 3:1 HRB™ (High Resolution Blend) is a mixture of agarose formulated to provide high resolution of small nucleic acids and PCR products. This product is specially designed for analysis of DNA fragments less than 1000 bp. Agarose 3:1 HRB™ displays precise banding patterns without smearing or high background fluorescence. The high gel strength allows for easy to handle gels that are optimal for blotting applications.
  • High resolution blend
  • Optimal for small nucleic acid fragments and PCR products
  • Easy to handle due to high gel strength
  • Nuclease- and protease-free