Hand cleansers, Estesol® mild wash
Leverandør: Deb Stoko
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EVON22309EA 53.8 DKK
EVON22309
Hand cleansers, Estesol® mild wash
Estesol® Mild Wash is a creamy, special skin cleanser featuring mild cleansing power and good skin compatibility. It is designed for all types of light contamination which could otherwise only be removed by using scrubbing agents and solvents. Water-soluble.
  • Very good skin compatibility, can be used all over the body, and even on sensitive skin
  • Does not contain any perfumes or colourants: Can also be used in food industry applications
  • With skin smoothing conditioner: Improved skin feel after washing coupled with caring effect on the skin
Certificeringer: EU Ecolabel