Pellet presses, PP 40
Leverandør: Retsch
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RETS20.750.0002EA 335560 DKK
RETS20.750.0002 RETS20.750.0003 RETS20.750.0004 RETS20.750.0005
Pellet presses, PP 40
High-quality pellets are a precondition for X-ray fluorescence (XRF) analysis.
  • Stong pellets with a smooth surface
  • Determines the time of build-up, holding and release of force during pressing
  • Reduced tensions of the sample fo difficult materials to be pressed
Designed for the preparation of pellets for spectral analyses.
Power requirements: 220 - 230 V/50 - 60 Hz.