Pellet press, PP 25
Leverandør: Retsch
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RETS20.750.0006EA 80680 DKK
RETS20.750.0006
Pellet press, PP 25
The PP 25 is a benchtop unit with simple and safe operation.
  • Pellets produced are of high quality and are characterized by their high degree of stability
  • Piston pressure can be read off from the clearly visible manometer scale
  • Dies available in several diameters and can be evacuated completely
High-quality pellets are an important precondition for reliable and meaningful XRF analysis.