Cold resistant gloves, XRT Insulator
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
112-2846EA 127 DKK
MARGM9999351 MARGM9999352 MARGM9999353 MARGM9999354
Cold resistant gloves, XRT Insulator
These synthetic leather gloves with polyamide/Lycra® back provide protection against mechanical hazards and low temperatures down to −30 °C.
  • Elastic tightening feature at wrist (zigzag finish)
  • Good abrasion resistance
  • High flexibility and softness
  • Back in a high visibility colour for improved safety
Ideal for use in cold stores as well as for general use in all engineering, mechanics, transport, construction and outdoor applications in cold environments.
Certificeringer: EN 388 (2222), EN 511 (X20), Cat. II