Harnesses, Titan™
Leverandør: Honeywell Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Miller® Miller® Titan™
139-0232EA 485 DKK
139-0232 113-1269
Harnesses, Titan™
Faldbeskyttelse Faldseler
Non stretch harnesses for construction work. Available as a 1-point version with rear anchorage or as a 2-point version with front and rear anchorage.
  • Lightweight, durable polyester webbing for all-day comfort and dependability
  • Fully adjustable designs so one size fits all
  • Sliding back D-ring for a comfortable fit
  • Sub pelvic strap keeps the wearer upright
  • Chest strap (45 mm) with metal buckles
Certificeringer: EN 361