Trousers, Fristads® PR54-220, Design A, royal blue
Leverandør: Fristads Kansas Group
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Fristads®
113-02596EA 0 DKK
113-02596 113-02597 113-02630 113-02594 113-02595 113-02592 113-02593 113-02590 113-02591 113-00733 113-02637 113-02638 113-00732 113-02635 113-00735 113-00734 113-02636 113-02633 113-02634 113-00731 113-02631 113-02598 113-02632 113-00730 113-02599 113-00729 113-00726 113-00725 113-00728 113-02628 113-02629 113-00727 113-02640 113-02641 113-02648 113-00744 113-00743 113-00746 113-02646 113-00745 113-02647 113-02644 113-00740 113-02645 113-00742 113-02642 113-00741 113-02643 113-00737 113-00736 113-00739 113-02639 113-00738 113-00755 113-00754 113-00757 113-00756 113-00751 113-00750 113-00753 113-00752 113-00748 113-00747 113-00749 113-00760 113-00766 113-00765 113-00768 113-00801 113-00767 113-00800 113-00762 113-00761 113-00764 113-00763 113-00759 113-00758 113-00771 113-00770 113-00810 113-00777 113-00776 113-00779 113-00812 113-00811 113-00778 113-00773 113-00772 113-00775 113-00774 113-00807 113-00806 113-00809 113-00808 113-00803 113-00802 113-00769 113-00805 113-00804 113-00780 113-00782 113-00781 113-00788 113-02604 113-00821 113-00787 113-00820 113-02605 113-02602 113-00823 113-02603 113-00789 113-00822 113-00784 113-02600 113-00783 113-02601 113-00786 113-00785 113-00818 113-00817 113-00819 113-00814 113-00813 113-00816 113-00815 113-00791 113-00790 113-00793 113-00792 113-00711 113-00799 113-02615 113-00798 113-02616 113-00710 113-00713 113-02613 113-02614 113-00712 113-02611 113-02578 113-00795 113-00794 113-02579 113-02612 113-00797 113-02610 113-00796 113-02577 113-00708 113-00707 113-00709 113-02608 113-00825 113-02609 113-00824 113-02606 113-00706 113-02607 113-02585 113-02586 113-02583 113-02584 113-02581 113-02582 113-02580 113-02626 113-00722 113-00721 113-02627 113-02624 113-00724 113-00723 113-02625 113-02622 113-02589 113-02623 113-00720 113-02587 113-02620 113-02588 113-02621 113-00719 113-00718 113-02619 113-00715 113-00714 113-02617 113-00717 113-00716 113-02618
Trousers, Fristads® PR54-220, Design A, royal blue
Arbejdstøj
Thanks to their durability and comfort, these trousers are perfectly suitable for use in garages as well as in industrial and service companies. The material is made of 65% polyester and 35% cotton. The inside is brushed and provides a comfortable cotton feel. The outside is very hardwearing and does not bleach, even after countless washes.
  • Two front and back pockets
  • Two leg pockets - one with flap, telephone pocket, pen pocket, tool bags
  • Adjustable waist, plastic snap buttons
  • Colour accents on pocket edges
Weight: 260 g/m²
Basic colour: royal blue