Reusable half masks, 7000 series, 3M™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MMMA7003EA 0 DKK
MMMA7003
Reusable half masks, 7000 series, 3M™
Åndedrætsværn Genanvendelige masker Halvmasker
7003 half mask is the larger sized mask in the 7000 half mask range.
  • With replaceable parts
  • Twin filter design provides low breathing resistance
  • Textured nose cup reduces face piece slippage and skin irritation caused by perspiration build-up
  • Unique 3M™ bayonet filter fixing system
  • With cradle head harness and easy to fasten neck strap
Certificeringer: EN 140