Remote digital thermo-hygrometer,625
Leverandør: Testo
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-3019EA 2680 DKK
620-3019
Remote digital thermo-hygrometer,625
Hygrometre Digitale termo-hygrometre
This instrument measures and displays temperature, humidity and dew point in offices, storage buildings and greenhouses. The sensor can be placed remotely from the device using an optional connecting cable. Humidity and temperature can also be measured using wireless radio probes. A temperature and relative humidity measurement can be taken at the same time as a surface temperature measurement, for example, using a radio probe. The display can be switched between temperature, dew point temperature and relative humidity as required.
  • Back-lit LCD
  • Minimum/maximum value display
  • "HOLD" function to freeze the displayed value
Leveringsinformation: Supplied with batteries and calibration protocole.