Chlortetracyclin, HCl
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
EHERC11509100EA
758
DKK
EHERC11509100
Chlortetracyclin, HCl
Chlortetracyclin, HCl
Formel:
C₂₂H₂₃ClN₂O₈·HCl Molekylvægt: 515,34 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD26142699 CAS-nummer: 64-72-2 EINECS: 200-591-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...