VWR®, Chemical Resistant Trousers
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
113-1959EA 328 DKK
113-1959 113-6171 113-6170 113-6177 113-6133 113-6176 113-6132 113-6135 113-6179 113-6178 113-6134 113-6173 113-6172 113-6175 113-6131 113-6130 113-6174 113-1963 113-6137 113-1962 113-6136 113-1961 113-1960 113-1967 113-1966 113-1965 113-1964 113-1969 113-1968 113-8045 113-1970 113-1974 113-1973 113-1972 113-1971 113-1978 113-1977 113-1976 113-1975 113-6119 113-6118 113-6159 113-6117 113-6160 113-6122 113-6166 113-6165 113-6121 113-6124 113-6168 113-6123 113-6167 113-6162 113-6161 113-6164 113-6120 113-6163 113-6129 113-5999 113-6126 113-6169 113-6125 113-6128 113-6127
VWR®, Chemical Resistant Trousers
Arbejdstøj
These trousers are manufactured from 99% polyester fibres and 1% Hydro-Tec fibres. The special fabric, with Hydro-Tec nanotechnology finish, makes the trousers extremly durable and provides anti-static features as well as protection against dirt, oil, chemicals and water.
  • Excellent protection against chemical splashes
  • Two side and two back pockets, all covered by a flap
  • Elasticated waistband which can be adjusted for improved wearer comfort
  • Closure: Metal press stud with zip closure
Ideal for wearing in combination with the chemical resistant VWR® coats.
Colour: White, grey or blue
Density: 275 g/m²
Certificeringer: EN 13034:2005 (type 6), EN 1149-5; complies with Oeko-Tex® Standard 100