Goggles, uvex 9305
Leverandør: Uvex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
uvex 9305
UVEX9305.714EA 252 DKK
UVEX9305.714
Goggles, uvex 9305
Beskyttelsesbriller
Classic safety goggles with acetate lens.
  • Effective ventilation system
  • Adjustable textile headband
  • Mechanical strength: F –Low energy impact protection (45 m/s)
Certificeringer: EN 166, EN 170