Earmuffs, helmet-mounted, uvex 3H
Leverandør: Uvex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
UVEXUVEX.3H
Earmuffs, helmet-mounted, uvex 3H
Dielectric helmet-mounted earmuffs for safety helmets designed to follow the demands of weight balance when ear muff is in position on the helmet.
  • uvex 3H cap attachment comes with adapter to suit safety helmets with 25 mm slots
  • Adapters also available for 30 mm slot helmets (sold separately)
  • Uvex ultra soft cushion technology
  • Low clamping force of 12,5 Newtons for high wearer comfort
Suitable for use in workplace environments with exposure to electrical voltage.
SNR: 32 dB
Weight: 245 g
Certificeringer: EN 352-1; AS/NZS 1270