Colony count pen
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
710-0902EA 107 DKK
PBIB89749 PBIB89748
Colony count pen
Pen style electronic colony counter for rapid and accurate counts.
  • Audible alarm for end of count with reset button
  • Lightweight and ergonomic shape to make heavy workload operation easy
  • Once the counting is complete, the pen can record the counts
Ensuring precise counts, the colony counting pen can be reset at any time by pressing the white button or turning off the pen.