Ethalfluralin

Leverandør: EHRENSTORFER GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

EHERC13220000EA 1260 DKK
EHERC13220000
Ethalfluralin
Ethalfluralin
Formel: C₁₃H₁₄F₃N₃O₄
Molekylvægt: 333,27 g/mol
CAS-nummer: 55283-68-6
EINECS: 259-564-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR