Propachlor
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC16380000EA
1210
DKK
EHERC16380000
Propachlor
Propachlor
Formel:
C₁₁H₁₄ClNO Molekylvægt: 211,69 g/mol |
CAS-nummer:
1918-16-7 EINECS: 217-638-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...