Micro magnet for glass pumps, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BOLA
BOHLG876-14EA 1580 DKK
BOHLG876-14
Micro magnet for glass pumps, BOLA
Magneter
Made of PTFE, the magnet can be adjusted in height by means of the integrated screw. The strong magnet provides good power transmission. Suitable for all common laboratory glass pumps, e.g. from company NORMAG®.
  • Chemical resistance
  • Temperature resistance from +0 to +150 °C