Integrated ultrasonic cleaning systems, IC-Series
Leverandør: Branson
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BRANCPN-908-016EA 37080 DKK
BRANCPN-908-016 BRANCPN-908-026 BRANCPN-908-015 BRANCPN-908-025
Integrated ultrasonic cleaning systems, IC-Series
Integrated ultrasonic cleaning systems offer a full range of features to meet most precision cleaning requirements.
  • Microprocessor-controlled for precise process management
  • Digitally controlled temperature to 69 °C
  • Digital process timer from 1 to 99 minutes
  • High visibility digital display of time and temperature
It is available in two frequencies, 25/40 kHz. Generally, 25 kHz is preferred for large massive parts or parts which are more densely packed. The higher 40kHz frequency is often used for smaller, more intricate parts or components with complex geometries.
Certificeringer: Meets CSA and UL requirements.