Hotplates, Undergrad UC150 and US150
Leverandør: Cole Parmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
453-0143EA 3790 DKK
STUAUS150 STUAUC150
Hotplates, Undergrad UC150 and US150
Compact, general purpose hotplates designed for safety and performance. The hotplate has microprocessor control and an innovative LED based true °C temperature setting system rather than a scale. However, to accurately control sample temperature (from 50 to 200 °C) the unit can be used in conjunction with the SCT1 digital contact thermometer to accurately monitor sample temperature rather than plate temperature. Both models have an integral fitting for a retort rod.
  • Choice of top plate: Robust coated aluminium or chemically resistant ceramic
  • Flashing 'hot' warning light whenever the plate temperature is above 50 °C, is mains independent so stays on even when the unit is unplugged
  • Accurate temperature control with LED temperature indicator
  • Compact, space saving unit, can be stored on its side to maximise storage space
  • Designed with recess at the base for a retort stand to slide underneath unit, saves space and time when setting up
Model UC150 has a glass ceramic top which has excellent chemical resistance. The surface is easy to clean and allows high plate temperatures for faster heating.
Model US150 has a robust aluminium/silicon alloy top plate for excellent heat transmission. The top plate has a thin ceramic coating for added chemical resistance. A 700 W element gives rapid heating and ensures even temperature distribution across the whole surface of the plate.
Certificeringer: IP protection class according to DIN EN 60529: IP 32
Advarsel: * RT= Ambient