Infectious disease test
Leverandør: SureScreen Diagnostics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-1351EA 473 DKK
SUREMALBC
Infectious disease test
This infectious disease test is a rapid chromatographic immunoassay for the qualitative detection of circulating plasmodium falciparum.
  • Attractive cassette format and attractive design
  • High performance immunoassay technology
  • Best technology with full technical support
Used in whole blood to detect HRP-2.