VWR® Traceable®, Infrared Thermometer gun with Laser Beam
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR® Traceable®
620-2101EA 0 DKK
620-2101
VWR® Traceable®, Infrared Thermometer gun with Laser Beam
Termometre Digital termometre
This lightweight unit takes readings in liquids and solids in one second. The laser beam enables perfect temperature target sighting in petroleum, electronics, plants and outdoors. The thermometer also provides average and differential readings.
  • Field of view is 8:1
  • Adjustable emissivity from 0,3 to 1
  • Backlight, data HOLD function, low battery indicator, °C/°F button
  • Maximum/minimum memory that can store up to nine values
  • High/low audible alarm
Certificeringer: Complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Leveringsinformation: Supplied with wrist strap, soft-sided carrying case, 9 volt battery and Traceable® certificate of calibration.