VWR® LP series, Precision balances
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
611-2316EA 4140 DKK
611-2316 611-2315 611-3363 611-2314 611-2313 611-2312 611-3349
VWR® LP series, Precision balances
Vægte
Rugged, economical, balances that are ideal for educational establishments and laboratory use. A series that ranges from simple weighing to operation with automated internal calibration and with GLP date/time functionality. All models have a stainless steel top pan.
  • Selectable filter levels and measuring units
  • Choice of simple LCD or large graphic display (G) with added functionality for GLP and formulation
Note: Balances are now supplied without an internal battery to meet recent transport regulations. An optional external rechargeable battery (611-8684) may be purchased for portable operation of models shown with **.
User can choose three options for models with internal calibration (denoted by 'i' at end of the model name):
Balance will calibrate using the internal weight by pressing CAL button
Balance will automatically recalibrate when ambient temperature changes and/or after a certain time has passed (Incal+)
The user can choose to calibrate the balance with an external weight.
Certificeringer: EN 61326-1:2013, EN 61010-1: 2010, EN 61010-2-010:2014, EN 50581:2012.
Bestillingsoplysninger: For details of the full range of accessories including printer, software, external keypad and PC connection cable please visit the website or contact your local Avantor sales office. These balances are also available with a calibration certificate. Please contact your local Avantor sales office for details of this option.
Leveringsinformation: Supplied with plastic in use protective cover.
Advarsel: Tablet Systems are not currently accredited for use with verified/EC-type approved balances.
Tablet Systems are not suitable for LPxG models (large graphic display precision balances).
Models with a large graphic display are denoted by G in the model name. Similarly models with a W or C in the model name have a three-door or a circular windshield, D indicates dual range and i shows internal calibration. FC models are supplied with a factory calibration certificate. See vwr.com for details of the full range of models.