Aluminum standard for AAS, analytical standard, traceable to BAM, Supelco®, Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
06154-500MLEA 569 DKK
06154-500ML
Aluminum standard for AAS, analytical standard, traceable to BAM, Supelco®, Merck
Aluminium standard solution, 1000 mg/L Al in 0,05M hydrochloric acid (from AlCl₃) standard for AAS