Rack for slide trays
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
631-0964EA 0 DKK
VWRI631-0964
Rack for slide trays
Sturdy rack designed to hold five slide trays.
  • Matching clasps area for identification labels
  • Can be placed in refrigerators
  • Specifically used for cold storage of samples
Suitable for applications in microbiology, histology and pathology.