Racks for chest freezers
Leverandør: Heathrow Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HEATHS2862AEEA 1660 DKK
HEATHS2862AE
Racks for chest freezers
Racks, made of corrosion resistant stainless steel.
  • Compatible with major brands of chest freezers
  • Robust mounting brackets assure handles stay securely in place for easy use
Offering a variety of racks to safely store samples in your freezer. Racks store boxes up to a 135 x 135 mm footprint and accommodate 2" tall boxes (up to 53 mm).