Stopcocks for desiccator, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DURAN®
467-0133EA 428 DKK
467-0133 467-0079 467-1256 467-0080
Stopcocks for desiccator, DURAN®
Stophaner
DURAN® borosilicate glass 3.3, clear, with PTFE needle valve.