4-Arsanilicsyre
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
EHERC10300250EA
727
DKK
EHERC10300250
4-Arsanilicsyre
4-Arsanilicsyre
Formel:
C₆H₈AsNO₃ Molekylvægt: 217,06 g/mol |
MDL Number:
MFCD00007819 CAS-nummer: 98-50-0 EINECS: 202-674-3 UN nummer: 3465 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...