Amersham ECL™ gel system
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
28-9906-08EA 0 DKK
GEHE28-9906-08
Amersham ECL™ gel system
Horizontal mini gel electrophoresis system for high quality protein electrophoresis. Designed to make protein PAGE as simple as agarose gel DNA electrophoresis.
  • High lot-to-lot consistency of gels ensures reproducible, high-quality results
  • Easier sample application and reduced buffer consumption than conventional vertical systems, plus no risk of leakage
  • Economic - same ready-to-run gels used for native and denaturing conditions, together with long shelf life, minimise risk of scrapping
  • Robust gels for convenient handling in downstream detection steps
  • Everything required to access and cut the gel is included in the Amersham ECL™ Gel cassette
Leveringsinformation: Complete system includes gel box, precast gels, and premixed running buffer for polyacrylamide gel electrophoresis (PAGE). Each component must be ordered separately.