Natriumhydroxid 25%, EMPROVE® bio for biopharmaceutical production low iron, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
4.80659.9025EA 0 DKK
4.80659.9025 4.80659.9950 4.80659.1000
Natriumhydroxid 25%, EMPROVE® bio for biopharmaceutical production low iron, SAFC®
Sodium hydroxide
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: 119…135 °C (1013 hPa)
Smeltepunkt: 9…10 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8,II