MediLine laboratory refrigerator, with TC 02 command
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LIEBLGKV6410EA 13670 DKK
LIEBLGKV6410 LIEBLGKV5710
MediLine laboratory refrigerator, with TC 02 command
This MedilLne laboratory refrigerator line includes TC 02 command, which consists of an electronic circuit control, located behind the front panel or in a PVC box in the back of the refrigerator with a power reserve lead-acid battery 12V / 1.3Ah), a regulator and an electronic circuit with buttons / lights on the front panel. This special feature ensures the proper functioning of the refrigerator, the regulation of the internal temperature and warns users of any dysfunction.
  • Temperature regulation
  • Three temperature alarms registration, with date and duration
  • Open door warning
Temperature range: 0 °C to +15 °C