Stopcocks for burettes, straight, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DURAN®
SCOT286449405EA 8130 DKK
SCOT286449405
Stopcocks for burettes, straight, DURAN®
Stophaner
DURAN®, borosilicate glass 3.3, clear, grounded and polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.