Wipes for critical tasks, TuffStuff™
Leverandør: Contec
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TuffStuff™
CONTC1C1217QEA 0 DKK
CONTC1C1217Q CONTTSWC0917 CONTTSWC1217
Wipes for critical tasks, TuffStuff™
Papirservietter Engangsservietter og - håndklæder Papirservietter til følsomme opgaver
These heavy duty non woven wipes are made from polyester/cellulose fabric. They are hydroentangled for durability and cleanliness.
  • Hydroentangled non woven wipes are strong and durable without the use of binders
  • Offer strength wet or dry, no residues or binders left behind
  • Highly sorbent without the use of surfactants
  • Multiple wipe sizes allow optimum selection for each job
Ideal for tough cleaning and maintenance tasks, TuffStuff heavy duty wipes are excellent for cleaning up soil, oils, grime, solvents and lubricants, as well as applying solvents and thinners. They are very effective in applications such as surface preparations for coatings, machine operations and general maintenance.
Recommended for ISO Class 6 - 8 environments.